Assay Details
Target Gene Details
Entrez Gene ID: | 9056 |
Gene Name: | solute carrier family 7 member 7 |
Gene Aliases: |
LAT3, LPI, MOP-2, Y+LAT1, y+LAT-1 |
Location: |
Chr.14:22773222-22819811 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A7 | NM_001126105.2 | NP_001119577.1 | ||
NM_001126106.2 | NP_001119578.1 | |||
NR_040448.1 | ||||
XM_006720302.1 | XP_006720365.1 | |||
XM_011537298.2 | XP_011535600.1 | |||
XM_011537299.1 | XP_011535601.1 | |||
AB011263.1 | BAB11849.1 | |||
AB020532.1 | BAA87623.1 | |||
AB209591.1 | BAD92828.1 | |||
AF092032.1 | AAC83706.1 | |||
AJ130718.1 | CAA10198.1 | |||
AK025377.1 | ||||
AK301219.1 | ||||
AK314351.1 | ||||
AL365451.1 | ||||
BC003062.2 | AAH03062.1 | |||
BC010107.2 | AAH10107.1 | |||
BX161519.1 | CAD61952.1 | |||
BX248291.1 | CAD62619.1 | |||
KU178460.1 | ||||
KU178461.1 | ||||
KU178462.1 | ||||
Y18474.2 | CAB40136.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522410 | Chr.14:22788919 - 22801387 on Build GRCh38 | Loss | SLC7A7 |
dgv1836n100 | Chr.14:22676576 - 22803700 on Build GRCh38 | Gain | OXA1L SLC7A7 |
nsv563998 | Chr.14:22414183 - 22879743 on Build GRCh38 | Gain | TRDV2 OXA1L TRAC TRDV3 MRPL52 LOC105370399 TRAJ60 LRP10 SLC7A7 MMP14 TRAJ17 ABHD4 DAD1 |
esv3581073 | Chr.14:22794871 - 22811943 on Build GRCh38 | Loss | SLC7A7 |
esv3633763 | Chr.14:22686795 - 22843098 on Build GRCh38 | Gain | OXA1L SLC7A7 MMP14 MRPL52 |
nsv819358 | Chr.14:22775794 - 22811167 on Build GRCh38 | Loss | SLC7A7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |