Assay Details
Target Gene Details
Entrez Gene ID: | 55333 |
Gene Name: | synaptojanin 2 binding protein |
Gene Aliases: |
ARIP2, OMP25 |
Location: |
Chr.14:70366496-70417090 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYNJ2BP | NM_018373.2 | NP_060843.2 | ||
AK002133.1 | BAA92098.1 | |||
AK311719.1 | ||||
AY534623.1 | AAT01566.1 | |||
BC007704.2 | AAH07704.1 | |||
BP195583.1 |
Target Gene Details
Entrez Gene ID: | 100529257 |
Gene Name: | SYNJ2BP-COX16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:70325081-70417090 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SYNJ2BP-COX16 | NM_001202547.2 | NP_001189476.1 | ||
NM_001202548.2 | NP_001189477.1 | |||
NM_001202549.2 | NP_001189478.1 | |||
FY210787.1 | ||||
FY211163.1 | ||||
FY213292.1 | ||||
FY213836.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)