Assay Details
Target Gene Details
Entrez Gene ID: | 100750246 |
Gene Name: | HIF1A antisense RNA 1 |
Gene Aliases: |
5'aHIF-1A, 5'aHIF1alpha |
Location: |
Chr.14:61681041-61695823 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HIF1A-AS1 | NR_047116.1 | |||
CN270307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564884 | Chr.14:61667038 - 61740839 on Build GRCh38 | Loss | HIF1A-AS1 HIF1A |