Assay Details
Target Gene Details
Entrez Gene ID: | 9472 |
Gene Name: | A-kinase anchoring protein 6 |
Gene Aliases: |
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP |
Location: |
Chr.14:32329273-32833999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AKAP6 | NM_004274.4 | NP_004265.3 | ||
AB002309.2 | ||||
BC137232.1 | ||||
BC137233.1 | ||||
BC150185.1 | ||||
BC150288.1 | ||||
BC154413.1 | ||||
BP230135.1 | ||||
U17195.2 | AAA92354.2 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)