Assay Details
Target Gene Details
Entrez Gene ID: | 26512 |
Gene Name: | integrator complex subunit 6 |
Gene Aliases: |
DBI-1, DDX26, DDX26A, DICE1, HDB, INT6, Notchl2 |
Location: |
Chr.13:51353171-51453236 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
INTS6 | NM_001039937.1 | NP_001035026.1 | ||
NM_001306091.1 | NP_001293020.1 | |||
NM_012141.2 | NP_036273.1 | |||
XM_011535040.2 | XP_011533342.1 | |||
XM_011535041.1 | XP_011533343.1 | |||
XM_017020540.1 | XP_016876029.1 | |||
AF097645.1 | AAF03046.1 | |||
AF141326.1 | AAD39481.1 | |||
AK074946.1 | ||||
AK096696.1 | ||||
AK128795.1 | ||||
AK296801.1 | ||||
AK304218.1 | ||||
AL117626.1 | CAB56020.1 | |||
AL833524.1 | ||||
BC013358.2 | AAH13358.1 | |||
BC039829.1 | AAH39829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)