Assay Details
Target Gene Details
Entrez Gene ID: | 121340 |
Gene Name: | Sp7 transcription factor |
Gene Aliases: |
OI11, OI12, OSX, osterix |
Location: |
Chr.12:53326575-53344793 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SP7 | NM_001300837.1 | NP_001287766.1 | ||
XM_011537900.2 | XP_011536202.1 | |||
HY020346.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss | LOC283335 RARG IGFBP6 MFSD5 SPRYD3 PFDN5 ZNF740 C12orf10 TNS2 ITGB7 ESPL1 AAAS EIF4B SOAT2 CSAD MIR6757 SP7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |