Assay Details
Target Gene Details
Entrez Gene ID: | 41 |
Gene Name: | acid sensing ion channel subunit 1 |
Gene Aliases: |
ACCN2, ASIC, BNaC2 |
Location: |
Chr.12:50057637-50083622 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASIC1 | NM_001095.3 | NP_001086.2 | ||
NM_020039.3 | NP_064423.2 | |||
NR_046389.1 | ||||
XM_011538350.1 | XP_011536652.1 | |||
XM_011538351.2 | XP_011536653.1 | |||
XM_011538352.1 | XP_011536654.1 | |||
AK290010.1 | ||||
AK293960.1 | ||||
AK311047.1 | ||||
BC133707.1 | ||||
EU078959.1 | ||||
U78180.1 | AAB48980.1 | |||
U78181.1 | AAB48981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv558837 | Chr.12:49955982 - 50073986 on Build GRCh38 | Loss | AQP6 AQP2 RACGAP1 AQP5 ASIC1 LOC101927318 |
nsv952179 | Chr.12:50070218 - 50079817 on Build GRCh38 | Deletion | ASIC1 |
nsv558840 | Chr.12:50056193 - 50130181 on Build GRCh38 | Loss | CERS5 SMARCD1 ASIC1 COX14 GPD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |