Assay Details
Target Gene Details
Entrez Gene ID: | 80024 |
Gene Name: | solute carrier family 8 member B1 |
Gene Aliases: |
NCKX6, NCLX, SLC24A6 |
Location: |
Chr.12:113298766-113335142 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC8B1 | NM_024959.2 | NP_079235.2 | ||
XM_006719607.2 | XP_006719670.1 | |||
XM_011538749.2 | XP_011537051.1 | |||
XM_011538750.2 | XP_011537052.1 | |||
XM_011538752.2 | XP_011537054.1 | |||
AK025886.1 | BAB15271.1 | |||
AK094074.1 | ||||
AK095092.1 | ||||
AK293216.1 | ||||
AY601759.1 | AAT35807.1 | |||
AY601760.1 | AAT35808.1 | |||
BC098360.1 | AAH98360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss | SLC8B1 RASAL1 DTX1 SDSL PLBD2 DDX54 MIR6762 IQCD RITA1 CFAP73 SDS MIR7106 TPCN1 |
esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain | SLC8B1 RASAL1 SDSL PLBD2 DDX54 MIR6762 LHX5 IQCD RITA1 CFAP73 LHX5-AS1 SDS MIR7106 TPCN1 |
esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain | SLC8B1 RASAL1 DTX1 SDSL PLBD2 DDX54 MIR6762 LHX5 LINC01234 IQCD RBM19 RITA1 CFAP73 LHX5-AS1 SDS MIR7106 OAS2 TPCN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |