Assay Details
Target Gene Details
Entrez Gene ID: | 81575 |
Gene Name: | apolipoprotein L domain containing 1 |
Gene Aliases: |
VERGE |
Location: |
Chr.12:12725917-12791466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOLD1 | NM_001130415.1 | NP_001123887.1 | ||
BC042478.1 | AAH42478.1 | |||
DC413327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion | MIR613 CREBL2 HTR7P1 LOH12CR2 MANSC1 DDX47 APOLD1 GPRC5D GPRC5A GSG1 LOC105369662 DUSP16 LOC100506314 CDKN1B HEBP1 FAM234B LRP6 BORCS5 RPL13AP20 MIR614 GPR19 |
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss | MIR1244-4 LOC100506248 MIR613 CREBL2 BCL2L14 LOH12CR2 MANSC1 DDX47 APOLD1 ETV6 LOC105369662 DUSP16 CDKN1B LINC01252 LRP6 BORCS5 RPL13AP20 GPR19 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |