Assay Details
Target Gene Details
Entrez Gene ID: | 10106 |
Gene Name: | CTD small phosphatase 2 |
Gene Aliases: |
OS4, PSR2, SCP2 |
Location: |
Chr.12:57819927-57846964 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTDSP2 | NM_005730.3 | NP_005721.3 | ||
XM_005268556.1 | XP_005268613.1 | |||
AF000152.1 | AAB71816.1 | |||
AF022231.1 | AAD09331.1 | |||
AK226076.1 | ||||
AK291289.1 | ||||
AK310183.1 | ||||
AK310873.1 | ||||
AK311070.1 | ||||
AK312413.1 | ||||
AY279531.1 | AAP34399.1 | |||
BC065920.1 | AAH65920.1 | |||
CN313995.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv726 | Chr.12:57817396 - 57852112 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)