Assay Details
Target Gene Details
Entrez Gene ID: | 23085 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 1 |
Gene Aliases: |
Cast2, ELKS, ERC-1, RAB6IP2 |
Location: |
Chr.12:990010-1495933 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain | LOC100049716 NINJ2 WNK1 LOC105369595 RAD52 ERC1 B4GALNT3 |
nsv524382 | Chr.12:999564 - 1016309 on Build GRCh38 | Loss | ERC1 |
nsv556943 | Chr.12:997617 - 1111522 on Build GRCh38 | Loss | ERC1 |
nsv556942 | Chr.12:938365 - 1010792 on Build GRCh38 | Gain | RAD52 ERC1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |