Assay Details
Target Gene Details
Entrez Gene ID: | 84766 |
Gene Name: | calcium release activated channel regulator 2A |
Gene Aliases: |
EFCAB4B |
Location: |
Chr.12:3613366-3753216 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRACR2A | NM_001144958.1 | NP_001138430.1 | ||
XM_006719021.3 | XP_006719084.1 | |||
XM_011521034.2 | XP_011519336.1 | |||
XM_011521036.2 | XP_011519338.1 | |||
XM_011521037.2 | XP_011519339.1 | |||
XM_011521038.2 | XP_011519340.1 | |||
AK091124.1 | ||||
AK304017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv582 | Chr.12:3596830 - 3627639 on Build GRCh38 | Insertion | CRACR2A |
nsv468981 | Chr.12:3576597 - 3989342 on Build GRCh38 | Gain | PARP11 PRMT8 CRACR2A |
nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss | C12orf4 KCNA6 KCNA5 TSPAN9 CCND2-AS1 KCNA1 CCND2 PRMT8 LOC100128253 NDUFA9 FGF6 LOC101929584 FGF23 DYRK4 THCAT155 LOC101929549 PARP11 AKAP3 TIGAR RAD51AP1 LOC105369606 GALNT8 CRACR2A |
nsv1036828 | Chr.12:3607219 - 3846326 on Build GRCh38 | Gain | PARP11 CRACR2A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |