Assay Details
Target Gene Details
Entrez Gene ID: | 101928062 |
Gene Name: | long intergenic non-protein coding RNA 1481 |
Gene Aliases: |
- |
Location: |
Chr.12:70222190-70243360 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC01481 | NR_120460.1 | |||
NR_120461.1 | ||||
DA349921.1 | ||||
DB462634.1 | ||||
HY002672.1 | ||||
HY013176.1 | ||||
HY048577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3629884 | Chr.12:70233284 - 70252849 on Build GRCh38 | Gain | CNOT2 LINC01481 |
nsv763 | Chr.12:70214096 - 70236518 on Build GRCh38 | Deletion | LINC01481 |
nsv559237 | Chr.12:70139848 - 70233539 on Build GRCh38 | Loss | LINC01481 |