Assay Details
Target Gene Details
Entrez Gene ID: | 5939 |
Gene Name: | RNA binding motif single stranded interacting protein 2 |
Gene Aliases: |
SCR3 |
Location: |
Chr.12:56521812-56596196 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RBMS2 | NM_002898.3 | NP_002889.1 | ||
XM_005269059.4 | XP_005269116.1 | |||
XM_005269060.4 | XP_005269117.1 | |||
XM_005269061.3 | XP_005269118.1 | |||
XM_005269066.4 | XP_005269123.1 | |||
XM_006719541.3 | XP_006719604.1 | |||
XM_006719542.3 | XP_006719605.1 | |||
XM_006719543.3 | XP_006719606.1 | |||
XM_006719544.3 | XP_006719607.1 | |||
XM_011538638.2 | XP_011536940.1 | |||
XM_011538639.2 | XP_011536941.1 | |||
XM_011538640.2 | XP_011536942.1 | |||
AK293315.1 | ||||
AK299580.1 | ||||
AK304540.1 | ||||
AK308885.1 | ||||
BC027863.1 | AAH27863.1 | |||
BC072679.1 | AAH72679.1 | |||
D28483.1 | BAA05842.1 | |||
DC382164.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss | ARHGEF25 INHBE SDR9C7 R3HDM2 DDIT3 COQ10A INHBC SHMT2 MIR616 STAC3 LRP1 NABP2 BAZ2A APOF TAC3 SNORA105C KIF5A NAB2 TIMELESS NACA ANKRD52 OS9 SLC39A5 HSD17B6 B4GALNT1 STAT6 MYO1A STAT2 NXPH4 ZBTB39 ATP5B NDUFA4L2 MARS CNPY2 PIP4K2C SPRYD4 RNF41 SNORD59A CS PAN2 GLI1 LRP1-AS IL23A RBMS2 NEMP1 MIP SLC26A10 ARHGAP9 DCTN2 GLS2 DTX3 SNORD59B MIR1228 MBD6 LOC101927583 PRIM1 MIR6758 RDH16 GPR182 PTGES3 HBCBP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |