Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFYB | NM_006166.3 | NP_006157.1 | ||
XM_005268908.4 | XP_005268965.1 | |||
XM_005268909.4 | XP_005268966.1 | |||
XM_005268910.4 | XP_005268967.1 | |||
AK291934.1 | ||||
BC005316.1 | AAH05316.1 | |||
BC005317.1 | AAH05317.1 | |||
BC007035.1 | AAH07035.1 | |||
BG776259.1 | ||||
X59710.1 | CAA42230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3580446 | Chr.12:104129444 - 104138892 on Build GRCh38 | Loss | NFYB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |