Assay Details
Target Gene Details
Entrez Gene ID: | 81575 |
Gene Name: | apolipoprotein L domain containing 1 |
Gene Aliases: |
VERGE |
Location: |
Chr.12:12725917-12791466 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOLD1 | NM_001130415.1 | NP_001123887.1 | ||
BC042478.1 | AAH42478.1 | |||
DC413327.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion | DUSP16 CDKN1B LRP6 MIR613 BORCS5 GPRC5D GPRC5A CREBL2 FAM234B RPL13AP20 HTR7P1 MANSC1 DDX47 GSG1 MIR614 LOH12CR2 GPR19 LOC105369662 HEBP1 APOLD1 LOC100506314 |
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss | DUSP16 BCL2L14 CDKN1B MIR1244-4 LRP6 MIR613 BORCS5 LINC01252 CREBL2 RPL13AP20 MANSC1 DDX47 LOH12CR2 GPR19 LOC105369662 ETV6 LOC100506248 APOLD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |