Assay Details
Target Gene Details
Entrez Gene ID: | 144568 |
Gene Name: | alpha-2-macroglobulin like 1 |
Gene Aliases: |
CPAMD9 |
Location: |
Chr.12:8822472-8887202 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
A2ML1 | NM_144670.5 | NP_653271.2 | ||
XM_011520566.2 | XP_011518868.1 | |||
XM_011520567.2 | XP_011518869.1 | |||
XM_017018868.1 | XP_016874357.1 | |||
XM_017018869.1 | XP_016874358.1 | |||
XM_017018870.1 | XP_016874359.1 | |||
AK122624.1 | BAC85494.1 | |||
AK123601.1 | ||||
AK290355.1 | ||||
AL832139.1 | ||||
AL833479.1 | ||||
BX648231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain | LOC100506159 A2M-AS1 LOC400002 LOC101930452 LOC728715 CLECL1 LINC00612 KLRF2 LOC374443 A2M CLEC12A DDX12P CLEC2A CLEC4D CLEC4E M6PR MIR1244-3 AICDA CD69 KLRF1 CLEC2D RIMKLB CLEC6A CLEC2B KLRG1 PHC1 LOC105376678 A2MP1 A2ML1 PZP LOC105369728 OVOS MFAP5 LOC105369649 LOC642846 KLRB1 LOC101928030 LINC00987 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |