Assay Details
Target Gene Details
Entrez Gene ID: | 2004 |
Gene Name: | ELK3, ETS transcription factor |
Gene Aliases: |
ERP, NET, SAP-2, SAP2 |
Location: |
Chr.12:96194382-96269835 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ELK3 | NM_005230.3 | NP_005221.2 | ||
XM_006719275.3 | XP_006719338.1 | |||
AK312694.1 | ||||
BC017371.1 | AAH17371.1 | |||
DA754301.1 | ||||
Z36715.1 | CAA85309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv559842 | Chr.12:96194083 - 96197874 on Build GRCh38 | Gain | ELK3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |