Assay Details
Target Gene Details
Entrez Gene ID: | 51202 |
Gene Name: | DEAD-box helicase 47 |
Gene Aliases: |
E4-DBP, HQ0256, MSTP162, RRP3 |
Location: |
Chr.12:12813346-12829981 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX47 | NM_016355.3 | NP_057439.2 | ||
NM_201224.1 | NP_957518.1 | |||
AF078843.1 | AAF23354.1 | |||
AF190165.1 | AAQ13709.1 | |||
AJ276704.1 | CAC14786.1 | |||
AK054574.1 | BAB70762.1 | |||
AK222940.1 | BAD96660.1 | |||
AK225144.1 | ||||
AK302536.1 | ||||
AK310501.1 | ||||
AL136666.1 | CAB66601.1 | |||
AL833469.1 | ||||
BC009379.2 | AAH09379.2 | |||
BC068009.1 | AAH68009.1 | |||
EF036507.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion | HEBP1 APOLD1 DUSP16 MANSC1 LOH12CR2 LRP6 GPRC5D GPR19 HTR7P1 CREBL2 LOC105369662 FAM234B MIR614 LOC100506314 GPRC5A BORCS5 GSG1 MIR613 RPL13AP20 CDKN1B DDX47 |
nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss | BCL2L14 APOLD1 DUSP16 MANSC1 LOH12CR2 LRP6 GPR19 LOC100506248 CREBL2 LOC105369662 LINC01252 BORCS5 ETV6 MIR613 MIR1244-4 RPL13AP20 CDKN1B DDX47 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |