Assay Details
Target Gene Details
Entrez Gene ID: | 56262 |
Gene Name: | leucine rich repeat containing 8 family member A |
Gene Aliases: |
AGM5, LRRC8, SWELL1 |
Location: |
Chr.9:128882112-128918042 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LRRC8A | NM_001127244.1 | NP_001120716.1 | ||
NM_001127245.1 | NP_001120717.1 | |||
NM_019594.3 | NP_062540.2 | |||
XM_005252095.2 | XP_005252152.1 | |||
XM_005252096.3 | XP_005252153.1 | |||
XM_006717186.2 | XP_006717249.1 | |||
XM_011518863.1 | XP_011517165.1 | |||
XM_011518864.2 | XP_011517166.1 | |||
XM_011518865.2 | XP_011517167.1 | |||
AB037858.1 | ||||
AK024649.1 | ||||
AK289842.1 | ||||
BC051322.1 | AAH51322.1 | |||
BX376144.2 | ||||
DA243590.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)