Assay Details
Target Gene Details
Entrez Gene ID: | 26086 |
Gene Name: | G-protein signaling modulator 1 |
Gene Aliases: |
AGS3 |
Location: |
Chr.9:136327476-136359605 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GPSM1 | NM_001145638.2 | NP_001139110.2 | ||
NM_015597.5 | NP_056412.5 | |||
XM_011518498.2 | XP_011516800.1 | |||
BC009979.1 | AAH09979.1 | |||
BC017353.1 | AAH17353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1046074 | Chr.9:136228197 - 136393924 on Build GRCh38 | Gain | DNLZ CCDC187 CARD9 GPSM1 SNAPC4 LOC101928722 ESP33 DKFZP434A062 QSOX2 |
nsv825173 | Chr.9:135892081 - 136495763 on Build GRCh38 | Gain | GPSM1 SNAPC4 LOC101928722 UBAC1 ESP33 LHX3 DKFZP434A062 INPP5E LOC105376323 QSOX2 C9orf69 SEC16A C9orf163 DNLZ NOTCH1 NACC2 CCDC187 CARD9 SDCCAG3 LOC107987142 CAMSAP1 PMPCA |
nsv951202 | Chr.9:135770355 - 136558248 on Build GRCh38 | Deletion | MIR4673 GPSM1 SNAPC4 ESP33 KCNT1 INPP5E LOC105376323 NACC2 CCDC187 CAMSAP1 MIR4674 LOC101928722 UBAC1 LHX3 DKFZP434A062 QSOX2 C9orf69 SEC16A C9orf163 DNLZ NOTCH1 CARD9 SDCCAG3 LOC107987142 PMPCA NALT1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |