Assay Details
Target Gene Details
Entrez Gene ID: | 22949 |
Gene Name: | prostaglandin reductase 1 |
Gene Aliases: |
LTB4DH, PGR1, ZADH3 |
Location: |
Chr.9:111549722-111599855 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTGR1 | NM_001146108.1 | NP_001139580.1 | ||
NM_001146109.1 | NP_001139581.1 | |||
NM_012212.3 | NP_036344.2 | |||
XM_011518394.2 | XP_011516696.1 | |||
XM_011518395.2 | XP_011516697.1 | |||
XM_017014485.1 | XP_016869974.1 | |||
AK091948.1 | ||||
AK289597.1 | ||||
AK298379.1 | ||||
AK309866.1 | ||||
BC035228.2 | AAH35228.1 | |||
D49387.1 | BAA08382.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1054737 | Chr.9:111562245 - 111612449 on Build GRCh38 | Gain | ZNF483 PTGR1 LRRC37A5P |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |