Assay Details
Target Gene Details
Entrez Gene ID: | 169792 |
Gene Name: | GLIS family zinc finger 3 |
Gene Aliases: |
NDH, ZNF515 |
Location: |
Chr.9:3824127-4310694 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLIS3 | NM_001042413.1 | NP_001035878.1 | ||
NM_152629.3 | NP_689842.3 | |||
XM_005251386.4 | XP_005251443.1 | |||
XM_005251387.4 | XP_005251444.1 | |||
XM_005251388.4 | XP_005251445.1 | |||
XM_011517763.2 | XP_011516065.1 | |||
XM_011517764.2 | XP_011516066.1 | |||
XM_011517766.2 | XP_011516068.1 | |||
XM_011517767.2 | XP_011516069.1 | |||
XM_017014361.1 | XP_016869850.1 | |||
AB209404.1 | BAD92641.1 | |||
AK075059.1 | ||||
AK096318.1 | ||||
BC033899.2 | AAH33899.2 | |||
DQ438878.1 | ABE66435.1 | |||
KU178885.1 | ||||
KU178886.1 | ||||
KU178887.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1028973 | Chr.9:3789264 - 3856678 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)