Assay Details
Target Gene Details
Entrez Gene ID: | 25 |
Gene Name: | ABL proto-oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
ABL, JTK7, bcr/abl, c-ABL, c-ABL1, p150, v-abl |
Location: |
Chr.9:130713881-130887675 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABL1 | NM_007313.2 | NP_009297.2 | ||
AB209456.1 | BAD92693.1 | |||
AB209642.1 | BAD92879.1 | |||
AL707819.1 | ||||
BC117451.1 | ||||
M14753.1 | AAA51562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3621866 | Chr.9:130695968 - 131205700 on Build GRCh38 | Gain | FIBCD1 ABL1 QRFP LAMC3 EXOSC2 NUP214 AIF1L |
nsv831735 | Chr.9:130598942 - 130791581 on Build GRCh38 | Loss | FUBP3 ABL1 PRDM12 EXOSC2 MIR6856 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |