Assay Details
Target Gene Details
Entrez Gene ID: | 573 |
Gene Name: | BCL2 associated athanogene 1 |
Gene Aliases: |
BAG-1, HAP, RAP46 |
Location: |
Chr.9:33252471-33264761 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAG1 | NM_001172415.1 | NP_001165886.1 | ||
NM_004323.5 | NP_004314.5 | |||
AF022224.1 | AAC34258.1 | |||
AF116273.1 | AAD25045.1 | |||
AK093238.1 | ||||
AK096038.1 | ||||
AK222749.1 | BAD96469.1 | |||
AK316577.1 | ||||
BC001936.1 | ||||
BC014774.1 | ||||
KU177932.1 | ||||
KU177933.1 | ||||
U46917.1 | AAD11467.1 | |||
Z35491.1 | CAA84624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1023246 | Chr.9:33181636 - 33261063 on Build GRCh38 | Gain | BAG1 SPINK4 |
nsv519369 | Chr.9:33134619 - 33283163 on Build GRCh38 | Gain | CHMP5 B4GALT1 BAG1 SPINK4 B4GALT1-AS1 |
dgv7516n100 | Chr.9:33121909 - 33278846 on Build GRCh38 | Gain | CHMP5 B4GALT1 BAG1 SPINK4 B4GALT1-AS1 |
nsv614146 | Chr.9:33186158 - 33398608 on Build GRCh38 | Gain | CHMP5 NFX1 BAG1 AQP7 SPINK4 |
esv2762813 | Chr.9:33139254 - 33261075 on Build GRCh38 | Gain | B4GALT1 BAG1 SPINK4 B4GALT1-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |