Assay Details
Target Gene Details
Entrez Gene ID: | 549 |
Gene Name: | AU RNA binding protein/enoyl-CoA hydratase |
Gene Aliases: |
- |
Location: |
Chr.9:91213815-91361969 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AUH | NM_001306190.1 | NP_001293119.1 | ||
NM_001698.2 | NP_001689.1 | |||
XM_005252066.3 | XP_005252123.1 | |||
XM_005252067.4 | XP_005252124.1 | |||
XM_005252069.4 | XP_005252126.1 | |||
XM_006717150.3 | XP_006717213.1 | |||
XM_011518802.2 | XP_011517104.1 | |||
XM_017014849.1 | XP_016870338.1 | |||
XM_017014850.1 | XP_016870339.1 | |||
XM_017014851.1 | XP_016870340.1 | |||
BC020722.1 | AAH20722.1 | |||
BG706374.1 | ||||
X79888.1 | CAA56260.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3621048 | Chr.9:90655588 - 91365881 on Build GRCh38 | Gain | LINC00484 SYK LOC100129316 AUH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |