Assay Details
Target Gene Details
Entrez Gene ID: | 101927380 |
Gene Name: | C9orf41 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:74952965-74997382 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C9orf41-AS1 | NR_121183.1 | |||
BI552900.1 |