Assay Details
Target Gene Details
Entrez Gene ID: | 6505 |
Gene Name: | solute carrier family 1 member 1 |
Gene Aliases: |
DCBXA, EAAC1, EAAT3, SCZD18 |
Location: |
Chr.9:4490427-4587469 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC1A1 | NM_004170.5 | NP_004161.4 | ||
XM_011518007.1 | XP_011516309.1 | |||
XM_011518008.2 | XP_011516310.1 | |||
XM_011518009.2 | XP_011516311.1 | |||
XM_011518010.1 | XP_011516312.1 | |||
XM_017015042.1 | XP_016870531.1 | |||
XM_017015043.1 | XP_016870532.1 | |||
AB008536.1 | BAB83767.1 | |||
AK313013.1 | ||||
BC033040.1 | AAH33040.1 | |||
U03506.1 | AAA50430.1 | |||
U06469.1 | AAA53215.1 | |||
U08989.1 | AAA68628.1 |
Target Gene Details
Entrez Gene ID: | 55064 |
Gene Name: | spermatogenesis associated 6 like |
Gene Aliases: |
C9orf68, bA6J24.2 |
Location: |
Chr.9:4540092-4666674 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPATA6L | XM_017014882.1 | XP_016870371.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1022870 | Chr.9:4416046 - 4611901 on Build GRCh38 | Loss | SLC1A1 SPATA6L |
nsv8407 | Chr.9:4526442 - 4566030 on Build GRCh38 | Loss | SLC1A1 SPATA6L |
nsv1022493 | Chr.9:4490134 - 4604192 on Build GRCh38 | Loss | SLC1A1 SPATA6L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |