Assay Details
Target Gene Details
Entrez Gene ID: | 158234 |
Gene Name: | tRNA methyltransferase 10B |
Gene Aliases: |
RG9MTD3, bA3J10.9 |
Location: |
Chr.9:37753204-37780459 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831552 | Chr.9:37693652 - 37859980 on Build GRCh38 | Gain | TRMT10B DCAF10 EXOSC3 LOC105376037 FRMPD1 |
esv3891658 | Chr.9:37689626 - 37790130 on Build GRCh38 | Gain | TRMT10B EXOSC3 LOC105376037 FRMPD1 |
nsv469813 | Chr.9:37628200 - 37777281 on Build GRCh38 | Loss | RAB1C TRMT10B FRMPD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |