Assay Details
Target Gene Details
Entrez Gene ID: | 158293 |
Gene Name: | family with sequence similarity 120A opposite strand |
Gene Aliases: |
C9orf10OS |
Location: |
Chr.9:93446500-93453592 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM120AOS | NM_001322224.1 | NP_001309153.1 | ||
NM_198841.2 | NP_942138.2 | |||
NR_136229.1 | ||||
NR_136230.1 | ||||
NR_136231.1 | ||||
NR_136232.1 | ||||
NR_136233.1 | ||||
NR_136234.1 | ||||
NR_136235.1 | ||||
NR_136236.1 | ||||
NR_136237.1 | ||||
NR_136238.1 | ||||
AK056096.1 | ||||
AK309269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2672054 | Chr.9:93408253 - 93450030 on Build GRCh38 | Deletion | FAM120AOS |
esv3621099 | Chr.9:93408196 - 93450082 on Build GRCh38 | Loss | FAM120AOS |