Assay Details
Target Gene Details
Entrez Gene ID: | 51490 |
Gene Name: | SPOUT domain containing methyltransferase 1 |
Gene Aliases: |
C9orf114, CENP-32, HSPC109 |
Location: |
Chr.9:128819651-128833309 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPOUT1 | XM_017014804.1 | XP_016870293.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv6724 | Chr.9:128823206 - 128858415 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)