Assay Details
Target Gene Details
Entrez Gene ID: | 1056 |
Gene Name: | carboxyl ester lipase |
Gene Aliases: |
BAL, BSDL, BSSL, CELL, CEase, FAP, FAPP, LIPA, MODY8 |
Location: |
Chr.9:133061978-133071863 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEL | NM_001807.4 | NP_001798.2 | ||
AK299965.1 | ||||
BC042510.1 | AAH42510.1 | |||
M85201.1 | AAA52014.1 | |||
S79774.1 | AAB35488.2 | |||
X54457.1 | CAA38325.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1161913 | Chr.9:133059798 - 133090275 on Build GRCh38 | Duplication |
![]() ![]() ![]() |
nsv6746 | Chr.9:133044293 - 133068877 on Build GRCh38 | Insertion |
![]() ![]() ![]() ![]() |
nsv615596 | Chr.9:133030314 - 133080285 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv951795 | Chr.9:133060014 - 133070813 on Build GRCh38 | Deletion |
![]() ![]() |
esv3621931 | Chr.9:133063192 - 133080639 on Build GRCh38 | Loss |
![]() |
dgv12925n54 | Chr.9:133061291 - 133082065 on Build GRCh38 | Gain |
![]() ![]() |
nsv819112 | Chr.9:133058467 - 133082220 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2764128 | Chr.9:133061625 - 133082053 on Build GRCh38 | Gain+Loss |
![]() |
nsv831744 | Chr.9:132980050 - 133174043 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3573407 | Chr.9:133061226 - 133081633 on Build GRCh38 | Loss |
![]() ![]() |
dgv1242n67 | Chr.9:133061315 - 133084032 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2761264 | Chr.9:133054342 - 133082053 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv508564 | Chr.9:133013830 - 133073076 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
esv3576709 | Chr.9:133060063 - 133092673 on Build GRCh38 | Gain |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)