Assay Details
Target Gene Details
Entrez Gene ID: | 9329 |
Gene Name: | general transcription factor IIIC subunit 4 |
Gene Aliases: |
KAT12, TF3C-delta, TFIII90, TFIIIC290, TFIIIC90, TFIIICDELTA |
Location: |
Chr.9:132670035-132694955 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF3C4 | NM_012204.3 | NP_036336.2 | ||
NR_133925.1 | ||||
XM_017015317.1 | XP_016870806.1 | |||
XM_017015318.1 | XP_016870807.1 | |||
AF142328.1 | AAF05087.1 | |||
BC011619.2 | AAH11619.1 | |||
BC022787.1 | ||||
BC094774.1 | AAH94774.1 | |||
BC104755.1 | AAI04756.1 | |||
BC112245.1 | AAI12246.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |