Assay Details
Target Gene Details
Entrez Gene ID: | 23137 |
Gene Name: | structural maintenance of chromosomes 5 |
Gene Aliases: |
SMC5L1 |
Location: |
Chr.9:70258962-70354873 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMC5 | NM_015110.3 | NP_055925.2 | ||
XM_005251837.1 | XP_005251894.1 | |||
XM_005251838.1 | XP_005251895.1 | |||
XM_005251839.1 | XP_005251896.1 | |||
XM_017014507.1 | XP_016869996.1 | |||
XM_017014508.1 | XP_016869997.1 | |||
XM_017014509.1 | XP_016869998.1 | |||
AB011166.2 | ||||
AJ310550.1 | CAC39247.1 | |||
AK055825.1 | ||||
AK304133.1 | ||||
BC017666.1 | ||||
BC038225.1 | AAH38225.1 | |||
DB220819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1047862 | Chr.9:70131959 - 70523371 on Build GRCh38 | Gain | MAMDC2 MAMDC2-AS1 SMC5-AS1 KLF9 SMC5 |
nsv831615 | Chr.9:70237789 - 70404776 on Build GRCh38 | Gain | SMC5-AS1 KLF9 SMC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |