Assay Details
Target Gene Details
Entrez Gene ID: | 55003 |
Gene Name: | PAK1 interacting protein 1 |
Gene Aliases: |
MAK11, PIP1, WDR84, bA421M1.5, hPIP1 |
Location: |
Chr.6:10690865-10709782 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PAK1IP1 | NM_017906.2 | NP_060376.2 | ||
XM_005249204.2 | XP_005249261.1 | |||
XM_011514720.1 | XP_011513022.1 | |||
XM_011514721.1 | XP_011513023.1 | |||
AF283303.1 | AAK57477.1 | |||
AK000631.1 | BAA91296.1 | |||
AK225516.1 | ||||
BC010907.1 | AAH10907.1 | |||
KU178675.1 | ||||
KU178676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv966670 | Chr.6:10447438 - 10871656 on Build GRCh38 | Duplication | C6orf52 TMEM14B GCNT2 TMEM14C MIR5689HG MAK LOC101928106 PAK1IP1 |
nsv462631 | Chr.6:10449025 - 10839526 on Build GRCh38 | Gain | C6orf52 TMEM14B GCNT2 TMEM14C MIR5689HG MAK LOC101928106 PAK1IP1 |
nsv470800 | Chr.6:10538711 - 10863092 on Build GRCh38 | Gain | C6orf52 TMEM14B GCNT2 TMEM14C MAK PAK1IP1 |
nsv520853 | Chr.6:10671531 - 10724327 on Build GRCh38 | Loss | C6orf52 TMEM14C PAK1IP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |