Assay Details
Target Gene Details
Entrez Gene ID: | 262 |
Gene Name: | adenosylmethionine decarboxylase 1 |
Gene Aliases: |
ADOMETDC, AMD, SAMDC |
Location: |
Chr.6:110814621-110895713 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AMD1 | NM_001287215.1 | NP_001274144.1 | ||
AK304351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1032572 | Chr.6:110800096 - 111060820 on Build GRCh38 | Gain | AMD1 CDK19 GSTM2P1 GTF3C6 RPF2 |
nsv830773 | Chr.6:110813116 - 110985229 on Build GRCh38 | Loss | AMD1 CDK19 GTF3C6 RPF2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |