Assay Details
Target Gene Details
Entrez Gene ID: | 26002 |
Gene Name: | monooxygenase DBH like 1 |
Gene Aliases: |
MOX, PRO5780, dJ248E1.1 |
Location: |
Chr.6:132296055-132401534 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOXD1 | NM_015529.3 | NP_056344.2 | ||
XM_017010714.1 | XP_016866203.1 | |||
XM_017010715.1 | XP_016866204.1 | |||
AK074879.1 | BAC11263.1 | |||
AK226054.1 | ||||
AL080058.1 | CAB45692.1 | |||
AY007239.1 | AAG09636.1 | |||
AY359094.1 | AAQ89452.1 | |||
BC018756.1 | AAH18756.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)