Assay Details
Target Gene Details
Entrez Gene ID: | 29964 |
Gene Name: | prickle planar cell polarity protein 4 |
Gene Aliases: |
C6orf49, OBTP, OEBT, TOMM6 |
Location: |
Chr.6:41780762-41787372 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRICKLE4 | NM_013397.5 | NP_037529.3 | ||
BC110459.1 | AAI10460.1 | |||
DA875485.1 | ||||
DA957684.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |