Assay Details
Target Gene Details
Entrez Gene ID: | 221692 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
RPEL, RPEL1, dJ257A7.2 |
Location: |
Chr.6:12716635-13290484 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHACTR1 | NM_001242648.2 | NP_001229577.1 | ||
NM_001322308.1 | NP_001309237.1 | |||
NM_001322309.1 | NP_001309238.1 | |||
NM_001322310.1 | NP_001309239.1 | |||
NM_030948.3 | NP_112210.1 | |||
XM_005248934.2 | XP_005248991.1 | |||
XM_017010454.1 | XP_016865943.1 | |||
XM_017010455.1 | XP_016865944.1 | |||
XM_017010456.1 | XP_016865945.1 | |||
XM_017010457.1 | XP_016865946.1 | |||
XM_017010459.1 | XP_016865948.1 | |||
XM_017010460.1 | XP_016865949.1 | |||
XM_017010461.1 | XP_016865950.1 | |||
XM_017010462.1 | XP_016865951.1 | |||
AB051520.1 | BAB21824.1 | |||
AK090769.1 | ||||
AK290017.1 | ||||
AK295263.1 | ||||
BC101526.1 | AAI01527.1 | |||
BC101528.1 | AAI01529.1 | |||
CR746945.1 | ||||
DA253448.1 | ||||
DA330657.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)