Assay Details
Target Gene Details
Entrez Gene ID: | 64094 |
Gene Name: | SPARC related modular calcium binding 2 |
Gene Aliases: |
DTDP1, MST117, MSTP117, MSTP140, SMAP2, bA270C4A.1, bA37D8.1, dJ421D16.1 |
Location: |
Chr.6:168441151-168667994 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SMOC2 | NM_001166412.1 | NP_001159884.1 | ||
NM_022138.2 | NP_071421.1 | |||
XM_011536065.1 | XP_011534367.1 | |||
XM_011536066.1 | XP_011534368.1 | |||
AB014730.1 | BAB20267.1 | |||
AB014737.1 | BAB20274.1 | |||
AJ420521.1 | ||||
AK056700.1 | ||||
AK314613.1 | ||||
AL832303.1 | CAI46175.1 | |||
BC028420.1 | AAH28420.1 | |||
BC047583.1 | AAH47583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830868 | Chr.6:168333086 - 168526732 on Build GRCh38 | Gain | SMOC2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |