Assay Details
Target Gene Details
Entrez Gene ID: | 221656 |
Gene Name: | lysine demethylase 1B |
Gene Aliases: |
AOF1, C6orf193, LSD2, bA204B7.3, dJ298J15.2 |
Location: |
Chr.6:18155388-18223854 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KDM1B | NM_153042.3 | NP_694587.3 | ||
XM_005248925.1 | XP_005248982.1 | |||
XM_005248926.1 | XP_005248983.1 | |||
XM_005248928.1 | XP_005248985.1 | |||
XM_011514385.1 | XP_011512687.1 | |||
XM_011514386.1 | XP_011512688.1 | |||
XM_017010440.1 | XP_016865929.1 | |||
XM_017010441.1 | XP_016865930.1 | |||
XM_017010442.1 | XP_016865931.1 | |||
XM_017010443.1 | XP_016865932.1 | |||
XM_017010445.1 | XP_016865934.1 | |||
XM_017010446.1 | XP_016865935.1 | |||
AK125318.1 | BAC86124.1 | |||
BC113093.1 | ||||
DA099422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1034755 | Chr.6:18064320 - 18173781 on Build GRCh38 | Loss | TPMT KDM1B NHLRC1 |
nsv601050 | Chr.6:18134947 - 18221240 on Build GRCh38 | Gain | TPMT KDM1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |