Assay Details
Target Gene Details
Entrez Gene ID: | 51660 |
Gene Name: | mitochondrial pyruvate carrier 1 |
Gene Aliases: |
BRP44L, CGI-129, MPYCD, dJ68L15.3 |
Location: |
Chr.6:166364919-166383013 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MPC1 | NM_001270879.1 | NP_001257808.1 | ||
NM_016098.3 | NP_057182.1 | |||
NR_073093.1 | ||||
AF125101.1 | AAD39918.1 | |||
AF151887.1 | AAD34124.1 | |||
AK310934.1 | ||||
AK312201.1 | ||||
AL556427.3 | ||||
AL561589.3 | ||||
BC000810.1 | AAH00810.1 | |||
BF665465.1 | ||||
BG828174.1 | ||||
DC402302.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)