Assay Details
Target Gene Details
Entrez Gene ID: | 101926915 |
Gene Name: | uncharacterized LOC101926915 |
Gene Aliases: |
- |
Location: |
Chr.6:46276324-46365508 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101926915 | NR_125838.1 |
Target Gene Details
Entrez Gene ID: | 10231 |
Gene Name: | regulator of calcineurin 2 |
Gene Aliases: |
CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4 |
Location: |
Chr.6:46220730-46492067 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RCAN2 | NM_001251973.1 | NP_001238902.1 | ||
NM_001251974.1 | NP_001238903.1 | |||
NM_005822.3 | NP_005813.2 | |||
XM_011514226.1 | XP_011512528.1 | |||
AK090990.1 | ||||
AK312392.1 | ||||
AY034085.1 | AAK59805.1 | |||
AY034086.1 | AAK59806.1 | |||
BC038509.1 | AAH38509.1 | |||
D83407.1 | BAA11911.1 | |||
DA106611.1 | ||||
DA113879.1 | ||||
DA207634.1 | ||||
DA409014.1 | ||||
DB289594.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114266797] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |