Assay Details
Target Gene Details
Entrez Gene ID: | 54901 |
Gene Name: | CDK5 regulatory subunit associated protein 1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.6:20534457-21232404 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDKAL1 | NM_017774.3 | NP_060244.2 | ||
XM_011514718.1 | XP_011513020.1 | |||
XM_011514719.2 | XP_011513021.1 | |||
XM_017010986.1 | XP_016866475.1 | |||
XM_017010987.1 | XP_016866476.1 | |||
AK000349.1 | BAA91102.1 | |||
AK128546.1 | BAC87494.1 | |||
AK291735.1 | ||||
AK310219.1 | ||||
BC121020.1 | ||||
BC121021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1024963 | Chr.6:20474387 - 20862380 on Build GRCh38 | Gain | E2F3 CDKAL1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |