Assay Details
Target Gene Details
Entrez Gene ID: | 54516 |
Gene Name: | mitochondrial translational release factor 1 like |
Gene Aliases: |
HMRF1L, MRF1L, mtRF1a |
Location: |
Chr.6:152987349-153007301 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTRF1L | NM_001114184.2 | NP_001107656.1 | ||
NM_001301047.2 | NP_001287976.1 | |||
NM_001301870.1 | NP_001288799.1 | |||
NM_001301871.1 | NP_001288800.1 | |||
NM_019041.6 | NP_061914.3 | |||
NR_126056.1 | ||||
XM_017010959.1 | XP_016866448.1 | |||
XM_017010960.1 | XP_016866449.1 | |||
AK027684.1 | BAB55295.1 | |||
AK095240.1 | ||||
AK225321.1 | ||||
AK225970.1 | ||||
AK290662.1 | ||||
AK297671.1 | ||||
BC011873.2 | AAH11873.1 | |||
BC014428.1 | AAH14428.1 | |||
BC105973.1 | AAI05974.1 | |||
BX376520.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs144796751] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |