Assay Details
Target Gene Details
Entrez Gene ID: | 81847 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
- |
Location: |
Chr.6:127266340-127289893 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)