Assay Details
Target Gene Details
Entrez Gene ID: | 167681 |
Gene Name: | protease, serine 35 |
Gene Aliases: |
C6orf158, dJ223E3.1 |
Location: |
Chr.6:83512475-83525704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRSS35 | NM_001170423.1 | NP_001163894.1 | ||
NM_153362.2 | NP_699193.2 | |||
AK291928.1 | ||||
AY358661.1 | AAQ89024.1 | |||
BC037170.1 | AAH37170.1 | |||
DA251169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3540072 | Chr.6:83518814 - 83522953 on Build GRCh38 | Deletion | PRSS35 |
nsv518229 | Chr.6:83519395 - 83524112 on Build GRCh38 | Loss | PRSS35 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |