Assay Details
Target Gene Details
Entrez Gene ID: | 101060264 |
Gene Name: | FOXP4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41523895-41548621 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOXP4-AS1 | NR_126415.1 | |||
NR_126416.1 | ||||
NR_126417.1 | ||||
BF194903.1 | ||||
BG028463.1 | ||||
BQ082169.1 | ||||
BQ128057.1 | ||||
BQ581966.1 |