Assay Details
Target Gene Details
Entrez Gene ID: | 22875 |
Gene Name: | ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative) |
Gene Aliases: |
NPP4 |
Location: |
Chr.6:46129964-46146699 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENPP4 | NM_014936.4 | NP_055751.1 | ||
AK291276.1 | ||||
BC018054.1 | AAH18054.1 | |||
BI548305.1 | ||||
DA261977.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3576147 | Chr.6:46106919 - 46185586 on Build GRCh38 | Gain | ENPP4 CLIC5 ENPP5 |
nsv1022912 | Chr.6:46125672 - 46148712 on Build GRCh38 | Loss | ENPP4 CLIC5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |