Assay Details
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT3H | NM_001261823.1 | NP_001248752.1 | ||
NM_003599.3 | NP_003590.1 | |||
NM_181356.2 | NP_852001.1 | |||
XM_011514949.2 | XP_011513251.1 | |||
XM_011514950.1 | XP_011513252.1 | |||
XM_011514951.2 | XP_011513253.1 | |||
XM_011514952.2 | XP_011513254.1 | |||
XM_011514953.2 | XP_011513255.1 | |||
XM_011514954.2 | XP_011513256.1 | |||
XM_017011369.1 | XP_016866858.1 | |||
XM_017011370.1 | XP_016866859.1 | |||
XM_017011371.1 | XP_016866860.1 | |||
XM_017011372.1 | XP_016866861.1 | |||
XM_017011373.1 | XP_016866862.1 | |||
XM_017011374.1 | XP_016866863.1 | |||
AF069734.1 | AAC39904.1 | |||
AF073930.1 | AAC36098.1 | |||
AK303838.1 | ||||
AK313874.1 | ||||
BC035816.1 | AAH35816.1 | |||
BC050384.1 | AAH50384.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |